Tripropylene glycol, Thermo Scientific Chemicals
Tripropylene glycol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tripropylene glycol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
4 L
Catalog number 031798.AK
also known as 031798-AK
Price (USD)/ Each
26.20
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
26.20
Add to cart
Tripropylene glycol, Thermo Scientific Chemicals
Catalog number031798.AK
Price (USD)/ Each
26.20
-
Add to cart
Chemical Identifiers
CAS24800-44-0
IUPAC Name1-{[1-(1-hydroxypropoxy)propan-2-yl]oxy}propan-1-ol
Molecular FormulaC9H20O4
InChI KeySNAJJYBUNRAGEF-UHFFFAOYNA-N
SMILESCCC(O)OCC(C)OC(O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to faintly yellow
FormLiquid
Assay (GC)>/=99.0 %
Chloride content20 ppm max
Tripropylene glycol is used for textile soaps, lubricants, cutting oils and greases. It acts as an intermediate in the preparation of urethanes. It is also used as a solvent in coatings and inks. It is utilized as varnish removers, emulsifiers and hydraulic fluids. As an additive, it is used for carburetor fuels. It is involved as a cooling media as well as an active ingredient of plasticizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only