Thermo Scientific Chemicals

Tripropylene glycol, Thermo Scientific Chemicals

Catalog number: 031798.K7
4 L, Each
Thermo Scientific Chemicals

Tripropylene glycol, Thermo Scientific Chemicals

Catalog number: 031798.K7
4 L, Each
Quantity
Catalog number: 031798.K7
also known as 031798-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24800-44-0
IUPAC Name
1-{[1-(1-hydroxypropoxy)propan-2-yl]oxy}propan-1-ol
Molecular Formula
C9H20O4
InChI Key
SNAJJYBUNRAGEF-UHFFFAOYNA-N
SMILES
CCC(O)OCC(C)OC(O)CC
Appearance (Color)
Clear, colorless to faintly yellow
Form
Liquid
Assay (GC)
>/=99.0 %
Chloride content
20 ppm max

Description

Tripropylene glycol is used for textile soaps, lubricants, cutting oils and greases. It acts as an intermediate in the preparation of urethanes. It is also used as a solvent in coatings and inks. It is utilized as varnish removers, emulsifiers and hydraulic fluids. As an additive, it is used for carburetor fuels. It is involved as a cooling media as well as an active ingredient of plasticizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text