Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, ACS, 99.8-100.1% (Assay, dried basis), Thermo Scientific Chemicals

Catalog number: 031801.A1
1 kg, Each
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, ACS, 99.8-100.1% (Assay, dried basis), Thermo Scientific Chemicals

Catalog number: 031801.A1
1 kg, Each
Quantity
Catalog number: 031801.A1
also known as 031801-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-86-1
IUPAC Name
2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula
C4H11NO3
InChI Key
LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES
NC(CO)(CO)CO
Absorbance
Absorbance Passes Test
Assay from Suppliers CofA
99.8-100.1% (dry basis)
Residual water
≤2.0%
Heavy Metals
≤5ppm
Impurity content
Fe: ≤5ppm

Description

Tris(hydroxymethyl)aminomethane is used as a buffer in biochemistry and molecular biology laboratories. It is used as a primary standard to standardize acid solutions for chemical analysis. It finds application in cell membranes to increase permeability. As an alternative to sodium bicarbonate, it used in the treatment of metabolic acidosis. It acts as a precursor for the preparation of polymers, oxazolones and oxazolidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text