n-Undecane, 99%, Thermo Scientific Chemicals
n-Undecane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Undecane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 031807.18
also known as 031807-18
Price (USD)/ Each
52.65
Online exclusive
58.30 
Save 5.65 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
52.65
Online exclusive
58.30 
Save 5.65 (10%)
Add to cart
n-Undecane, 99%, Thermo Scientific Chemicals
Catalog number031807.18
Price (USD)/ Each
52.65
Online exclusive
58.30 
Save 5.65 (10%)
-
Add to cart
Chemical Identifiers
CAS1120-21-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Impurity contentHydrocarbons < C-11: ≤0.4% (GC)
Assay from Supplier's CofA≥98.5% (GC)
Impurity contentHydrocarbons > C-11: ≤0.6% (GC)
n-Undecane is used as a mild sex attractant for various types of moths and cockroaches. It is also used as an internal standard in the gas chromatographic analysis. Further, it is used as lubricants and lubricant additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only