Thermo Scientific Chemicals

n-Undecane, 99%, Thermo Scientific Chemicals

Catalog number: 031807.30
250 g, Each
Thermo Scientific Chemicals

n-Undecane, 99%, Thermo Scientific Chemicals

Catalog number: 031807.30
250 g, Each
Quantity
Catalog number: 031807.30
also known as 031807-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1120-21-4
Appearance (Color)
Clear colorless
Form
Liquid
Impurity content
Hydrocarbons < C-11: ≤0.4% (GC)
Assay from Supplier's CofA
≥98.5% (GC)
Impurity content
Hydrocarbons > C-11: ≤0.6% (GC)

Description

n-Undecane is used as a mild sex attractant for various types of moths and cockroaches. It is also used as an internal standard in the gas chromatographic analysis. Further, it is used as lubricants and lubricant additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text