Methylcyclohexane, 99+%, Thermo Scientific Chemicals
Methylcyclohexane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methylcyclohexane, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 031861.36
also known as 031861-36
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
Add to cart
Methylcyclohexane, 99+%, Thermo Scientific Chemicals
Catalog number031861.36
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
-
Add to cart
Chemical Identifiers
CAS108-87-2
IUPAC Namemethylcyclohexane
Molecular FormulaC7H14
InChI KeyUAEPNZWRGJTJPN-UHFFFAOYSA-N
SMILESCC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥99.0%
CommentClear colorless
FormLiquid
Methylcyclohexane is used as rubber, paint and varnish, used solvents, oils, such as solvent extraction, which can also be used for organic synthesis, Standards for the calibration of the thermometer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only