Thermo Scientific Chemicals

Morpholine, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 031984.36
500 g, Each
Thermo Scientific Chemicals

Morpholine, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 031984.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 031984.36
also known as 031984-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-91-8
IUPAC Name
morpholine
Molecular Formula
C4H9NO
InChI Key
YNAVUWVOSKDBBP-UHFFFAOYSA-N
SMILES
C1COCCN1
Color (APHA)
15 max
Comment
Boiling range 126 - 130 ?
Assay (Aqueous acid-base Titration)
>/= 99.0%

Description

Morpholine is used to make other chemicals, as a corrosion inhibitor, and in detergents. It is also used as solvent and optical brightener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text