Thermo Scientific Chemicals

Oleic acid, 99%, Thermo Scientific Chemicals

Catalog number: 031997.14
25 g, Each
Thermo Scientific Chemicals

Oleic acid, 99%, Thermo Scientific Chemicals

Catalog number: 031997.14
25 g, Each
Quantity
Catalog number: 031997.14
also known as 031997-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-80-1
IUPAC Name
(9E)-octadec-9-enoic acid
Molecular Formula
C18H34O2
InChI Key
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
SMILES
CCCCCCCC\C=C\CCCCCCCC(O)=O
Appearance (Color)
Colorless to pale yellow
Assay (GC)
≥98.5%
Impurity content
≤0.3% ( sum of Linoleic and Linolenic acids)
Form
Liquid

Description

Oleic acid is used as an emulsifying agent for facial creams and lotions, as an accelerator in the rubber industry, as an emollient, as a solubilizing agent in aerosol products and as a soldering flux in stained glass work for joining lead came. It is used in the preparation of oleates and lotions. It acts as intermediate for drilling and fracking fluids in the oil and gas industry. It finds application as an excipient in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text