Zinc protoporphyrin, 96%, Thermo Scientific Chemicals
Zinc protoporphyrin, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc protoporphyrin, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
10 mg
0.25 g
Catalog number 032068.MF
also known as 032068-MF
Price (USD)/ Each
146.00
-
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (USD)/ Each
146.00
Add to cart
Zinc protoporphyrin, 96%, Thermo Scientific Chemicals
Catalog number032068.MF
Price (USD)/ Each
146.00
-
Add to cart
Specifications
Chemical Name or MaterialZinc protoporphyrin
CAS15442-64-5
ColorPurple to Red
Recommended StorageAmbient temperatures
RTECS NumberZG9210300
View more
Zinc protoporphyrin-9 is a potent, selective heme oxygenase inhibitor, interleukin-1 antagonist, GCS inhibitor, and NOS1, NOS2, and NOS3 inhibitor. In olfactory neuronal cultures, this product has been used to block CO biosynthesis, which lowered cGMP concentrations. Zinc protoporphyrin-9 is an extremely useful tool to elucidate the role of CO in signal transduction and as a neurotransmitter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only