Thermo Scientific Chemicals

Gallium (III) Nitrate Hydrate, 99.9%, metals basis, Thermo Scientific Chemicals

Catalog number: 032116.06
5 g, Each
Thermo Scientific Chemicals

Gallium (III) Nitrate Hydrate, 99.9%, metals basis, Thermo Scientific Chemicals

Catalog number: 032116.06
5 g, Each
Quantity
Catalog number: 032116.06
also known as 032116-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69365-72-6
IUPAC Name
gallium(3+) trinitrate
Molecular Formula
GaN3O9
InChI Key
CHPZKNULDCNCBW-UHFFFAOYSA-N
SMILES
[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Metal Impurities
0.1% max.

Description

Gallium(III) nitrate hydrate is used in the preparation of gallium nitride and other gallium compounds. It acts as a drug used to treat symptomatic hypercalcemia secondary to cancer. It finds application as an antineoplastic agent and also as an acting agent for the treatment of bone-resorptive diseases. It is involved to improve the cultural selectivity of medically significant fungi by suppression of contaminating bacterial flora.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text