Thermo Scientific Chemicals

Benzyldimethylhexadecylammonium chloride hydrate, 95%, Thermo Scientific Chemicals

Catalog number: 032377.18
50 g, Each
Thermo Scientific Chemicals

Benzyldimethylhexadecylammonium chloride hydrate, 95%, Thermo Scientific Chemicals

Catalog number: 032377.18
50 g, Each
Quantity
Catalog number: 032377.18
also known as 032377-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-18-9
IUPAC Name
benzyl(hexadecyl)dimethylazanium chloride
Molecular Formula
C25H46ClN
InChI Key
SXPWTBGAZSPLHA-UHFFFAOYSA-M
SMILES
[Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
Appearance (Color)
White to pale cream
Form
Powder
Assay from Supplier's CofA
≥94%

Description

Benzyldimethylhexadecylammonium chloride has been used in a study to assess the removal of cationic surfactants from aqueous solutions using activated carbon cloth (ACC). It has also been used in a study to investigate the prototropic behavior of the dye, 7-hydroxy-4-methylcoumarin (7H4MC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text