Thermo Scientific Chemicals

Dimethyl sulfoxide, Spectrophotometric Grade, 99.9+%, Thermo Scientific Chemicals

Catalog number: 032434.K7
4 L, Each
Thermo Scientific Chemicals

Dimethyl sulfoxide, Spectrophotometric Grade, 99.9+%, Thermo Scientific Chemicals

Catalog number: 032434.K7
4 L, Each
Quantity
Catalog number: 032434.K7
also known as 032434-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-68-5
IUPAC Name
methanesulfinylmethane
Molecular Formula
C2H6OS
InChI Key
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES
CS(C)=O
Assay (GC)
≥99.9%
Water Content (Karl Fischer Titration)
0.1%, Max
Optical Absorbance at
268 nm 1.00, max
Optical Absorbance at
275 nm 0.50, max
Optical Absorbance at
300 nm 0.20, max

Description

Dimethyl sulfoxide is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text