Thermo Scientific Chemicals

Tetrachloroethylene, Ultrapure, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals

Catalog number: 032437.K2
1 L, Each
Thermo Scientific Chemicals

Tetrachloroethylene, Ultrapure, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals

Catalog number: 032437.K2
1 L, Each
Quantity
Catalog number: 032437.K2
also known as 032437-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
127-18-4
IUPAC Name
tetrachloroethene
Molecular Formula
C2Cl4
InChI Key
CYTYCFOTNPOANT-UHFFFAOYSA-N
SMILES
ClC(Cl)=C(Cl)Cl
Residual water
0.05%
Residue on Evaporation
<0.0005%

Description

Tetrachloroethylene is used as solvent. It is involved in dry cleaning as well as in the degreasing of metal parts. It finds application as a neutrino detector. It is utilized as a paint stripper and a spot remover in consumer products. It acts as a precursor for the manufacture of other chemicals. It plays an important role in water repellants, paint removers, printing inks, glues, sealants, polishes and lubricants. As a film-forming electrolyte additive, it is used in the manufacture of lithium ion batteries and is actively involved as an extraction solvent in the determination of oil and grease in water by FT-IR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text