Thermo Scientific Chemicals

Dichloromethane, Spectrophotometric Grade, 99.7+%, stab. with amylene, Thermo Scientific Chemicals

Catalog number: 032440.M1
4 x 1 L, Each
Thermo Scientific Chemicals

Dichloromethane, Spectrophotometric Grade, 99.7+%, stab. with amylene, Thermo Scientific Chemicals

Catalog number: 032440.M1
4 x 1 L, Each
Quantity
Catalog number: 032440.M1
also known as 032440-M1
Price (USD)
Have Questions?

Chemical Identifiers

CAS
75-09-2
IUPAC Name
dichloromethane
Molecular Formula
CH2Cl2
InChI Key
YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES
ClCCl
Residual water
0.02%
Impurity content
Free halogens P.T.
Residue on Evaporation
3ppm

Description

Dichloromethane is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text