Thermo Scientific Chemicals

1-Butanol, Ultrapure, Spectrophotometric Grade, 99.0+%, Thermo Scientific Chemicals

Catalog number: 032443.K2
1 L, Each
Thermo Scientific Chemicals

1-Butanol, Ultrapure, Spectrophotometric Grade, 99.0+%, Thermo Scientific Chemicals

Catalog number: 032443.K2
1 L, Each
Quantity
Catalog number: 032443.K2
also known as 032443-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71-36-3
IUPAC Name
butan-1-ol
Molecular Formula
C4H10O
InChI Key
LRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILES
CCCCO
Residual water
≤0.1%
Residue on Evaporation
≤5ppm
Assay from Supplier's CofA
≥99.0%
Assay from Supplier's CofA
≥99.0%
Residual water
≤0.1%

Description

1-Butanol is primarily used as a solvent, as an intermediate in chemical synthesis, and as a fuel. It is also used as a paint thinner and a solvent in other coating applications where it is used as a relatively slow evaporating latent solvent in lacquers and ambient-cured enamels. It finds other uses such as a component of hydraulic and brake fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text