2-Methoxyethanol, Spectrophotometric Grade, 99% min, Thermo Scientific Chemicals
2-Methoxyethanol, Spectrophotometric Grade, 99% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxyethanol, Spectrophotometric Grade, 99% min, Thermo Scientific Chemicals

Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 032444.K2
also known as 032444-K2
Price (USD)/ Each
74.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
87.40 
Save 12.75 (15%)
-
Add to cart
Request bulk or custom format
Price (USD)/ Each
74.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
87.40 
Save 12.75 (15%)
Add to cart
2-Methoxyethanol, Spectrophotometric Grade, 99% min, Thermo Scientific Chemicals
Catalog number032444.K2
Price (USD)/ Each
74.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
87.40 
Save 12.75 (15%)
-
Add to cart
Chemical Identifiers
CAS109-86-4
IUPAC Name2-methoxyethan-1-ol
Molecular FormulaC3H8O2
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Color (APHA)≤10
Assay from Supplier's CofA≥99.0%
Assay (special)Titrable Acid: ≤0.002 meq/g
View more
2-Methoxyethanol is used as a solvent for many different purposes such as varnishes, dyes, and resins. It is also used as an additive in airplane deicing solutions. In organometallic chemistry it is commonly used for the synthesis ofVaska's complex and related compounds such as ruthenium hydridechlorocarbonyl tristriphenylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only