Acetone, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals
Acetone, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetone, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1 L
1 L
4 L
4 x 4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 032451.M1
also known as 032451-M1
Price (USD)/ Each
142.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
168.00 
Save 25.35 (15%)
-
Add to cart
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)/ Each
142.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
168.00 
Save 25.35 (15%)
Add to cart
Acetone, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals
Catalog number032451.M1
Price (USD)/ Each
142.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
168.00 
Save 25.35 (15%)
-
Add to cart
Chemical Identifiers
CAS67-64-1
IUPAC Namepropan-2-one
Molecular FormulaC3H6O
InChI KeyCSCPPACGZOOCGX-UHFFFAOYSA-N
SMILESCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
FormLiquid
Assay (GC)≥99.5%
Water Content (Karl Fischer Titration)0.50%, max
Optical Absorbance at330nm: 1.0AU, max
View more
Acetone is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only