1,2-Dichloroethane, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals
1,2-Dichloroethane, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Dichloroethane, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 032462.K7
also known as 032462-K7
Price (USD)/ Each
136.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
160.00 
Save 23.35 (15%)
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
136.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
160.00 
Save 23.35 (15%)
Add to cart
1,2-Dichloroethane, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals
Catalog number032462.K7
Price (USD)/ Each
136.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
160.00 
Save 23.35 (15%)
-
Add to cart
Chemical Identifiers
CAS107-06-2
IUPAC Name1,2-dichloroethane
Molecular FormulaC2H4Cl2
InChI KeyWSLDOOZREJYCGB-UHFFFAOYSA-N
SMILESClCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on Evaporation≤10 ppm
Appearance (Color)Clear colorless
Assay from Supplier's CofA≥99.0%
FormLiquid
Residual water≤0.02%
1,2-dichloroethane is used in the production of vinyl chloride which is used to make a variety of plastic and vinyl products including polyvinyl chloride (PVC) pipes, furniture and automobile upholstery, wall coverings, house wares, and automobile parts.1,2-Dichloroethane is also used generally as an intermediate for other organic chemical compounds and as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only