Tetrahydrofuran, Spectrophotometric Grade, 99.7+%, unstab., Thermo Scientific Chemicals
Tetrahydrofuran, Spectrophotometric Grade, 99.7+%, unstab., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydrofuran, Spectrophotometric Grade, 99.7+%, unstab., Thermo Scientific Chemicals

 
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
4 x 1 L
4 L
Catalog number 032468.K2
also known as 032468-K2
Price (USD)/ Each
80.65
Online exclusive
89.90 
Save 9.25 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
80.65
Online exclusive
89.90 
Save 9.25 (10%)
Add to cart
Tetrahydrofuran, Spectrophotometric Grade, 99.7+%, unstab., Thermo Scientific Chemicals
Catalog number032468.K2
Price (USD)/ Each
80.65
Online exclusive
89.90 
Save 9.25 (10%)
-
Add to cart
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Color (APHA)≤10
Assay (GC)≥99.7%
Optical Absorbance at212nm: ≤1.00
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only