Thermo Scientific Chemicals

Carbon disulfide, 99.9%, Thermo Scientific Chemicals

Catalog number: 032472.AK
250 mL, Each
Thermo Scientific Chemicals

Carbon disulfide, 99.9%, Thermo Scientific Chemicals

Catalog number: 032472.AK
250 mL, Each
Quantity
Catalog number: 032472.AK
also known as 032472-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-15-0
IUPAC Name
methanedithione
Molecular Formula
CS2
InChI Key
QGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILES
S=C=S
Assay (GC)
≥99.9%
Appearance (Color)
Colorless fo very faintly yellow
Form
Liquid

Description

Carbon disulfide is used in electronic vacuum tubes, in the manufacturing of carbon tetrachloride, rayon, soil disinfectants, as a solvent, manufacture of rayon, cellophane, and carbon tetrachloride, to produce rubber and chemicals. It is used in the manufacture of xanthates. It is also used as a reaction agent for the production of rubber vulcanization accelerators and soil sterilizers. It is an effective solvent for oils, waxes, sulfur and sulfur compounds. Carbon disulfide and carbon dioxide acts as resources for functional polymers. It is also useful for the synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text