Thermo Scientific Chemicals

Dicyclopentadiene, typically 95%, stab., Thermo Scientific Chemicals

Catalog number: 032546.30
250 g, Each
Thermo Scientific Chemicals

Dicyclopentadiene, typically 95%, stab., Thermo Scientific Chemicals

Catalog number: 032546.30
250 g, Each
Quantity
Catalog number: 032546.30
also known as 032546-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-73-6
IUPAC Name
tricyclo[5.2.1.0²,⁶]deca-3,8-diene
Molecular Formula
C10H12
InChI Key
HECLRDQVFMWTQS-UHFFFAOYNA-N
SMILES
C1C=CC2C3CC(C=C3)C12
Assay from Supplier's CofA
≥95.0%
Color (APHA)
≤20
Impurity content
Acyclic Dienes: ≤2.0%
Impurity content
BHT: ≤0.04%
Impurity content
C9, C10, C11 codimers: ≤2.0%

Description

Dicyclopentadiene is used in resins, particularly unsaturated polyester resins. It plays a major role in inks, adhesives and paints. It is also used as a monomer in polymerization reactions. It is a precursor for the preparation of endo-tetrahydrodicyclopentadiene, which reacts with aluminum chloride at higher temperature to give adamantine. It undergoes a retro-Diels-Alder reaction to yield cyclopentadiene, which acts as ligand in inorganic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text