Thermo Scientific Chemicals

Lithium borohydride, 95%, Thermo Scientific Chemicals

Catalog number: 032563.18
50 g, Each
Thermo Scientific Chemicals

Lithium borohydride, 95%, Thermo Scientific Chemicals

Catalog number: 032563.18
50 g, Each
Quantity
Catalog number: 032563.18
also known as 032563-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16949-15-8
IUPAC Name
lithium(1+) boranuide
Molecular Formula
BH4Li
InChI Key
UUKMSDRCXNLYOO-UHFFFAOYSA-N
SMILES
[Li+].[BH4-]
Assay (unspecified)
94.1% min

Description

Strong reducing agent. Used to reduce compounds containing ketones, aldehydes, ester carbonyls, acid chlorides, lactones, and epoxidesLithium borohydride is a versatile reducing agent for aldehydes, ketones, nitriles, primary amides, acid chlorides, lactones, epoxides, and esters. It is involved in the preparation of intercalation compounds and nanocomposites. It is used as a precursor for other borohydrides and acts as a catalyst in hydroboration reactions. It is known to be good reversible storage compound for anhydrous ammonia and hydrogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Strong reducing agent
  • Used to reduce compounds containing ketones, aldehydes, ester carbonyls, acid chlorides, lactones, and epoxides
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text