Thermo Scientific Chemicals

Potassium borohydride, 98%, Thermo Scientific Chemicals

Catalog number: 032570.09
10 g, Each
Thermo Scientific Chemicals

Potassium borohydride, 98%, Thermo Scientific Chemicals

Catalog number: 032570.09
10 g, Each
Quantity
Catalog number: 032570.09
also known as 032570-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13762-51-1
IUPAC Name
potassium boranuide
Molecular Formula
BH4K
InChI Key
ICRGAIPBTSPUEX-UHFFFAOYSA-N
SMILES
[BH4-].[K+]
Assay (unspecified)
97.5% min

Description

Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plasticsPotassium borohydride is used as a selective reducing agent for ketones, aldehydes and acid chlorides. It is used as a precursor in the production of chloramphenicol, vitamin A, thiopenicol, atropine and scopolamine. It is used to prepare high dispersive copper nanoparticles by the chemical reduction method using potassium borohydride as a reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text