Thermo Scientific Chemicals

Lithium metaborate, anhydrous, tech., Thermo Scientific Chemicals

Catalog number: 032576.22
100 g, Each
Thermo Scientific Chemicals

Lithium metaborate, anhydrous, tech., Thermo Scientific Chemicals

Catalog number: 032576.22
100 g, Each
Quantity
Catalog number: 032576.22
also known as 032576-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13453-69-5
IUPAC Name
tetraboron heptahydrate dilithium
Molecular Formula
B4H14Li2O7
InChI Key
DPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
Assay (unspecified)
95% min

Description

Lithium Metaborate is commonly used as a fused compound or a component for an assay. It has been used for the analysis of wild rice for silicon, calcium, magnesium and potassium. Lithium metaborate was used to fuse with solid samples for decomposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text