Thermo Scientific Chemicals

Dichlorophenylborane, 96%, Thermo Scientific Chemicals

Catalog number: 032583.06
5 g, Each
Thermo Scientific Chemicals

Dichlorophenylborane, 96%, Thermo Scientific Chemicals

Catalog number: 032583.06
5 g, Each
Quantity
Catalog number: 032583.06
also known as 032583-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
873-51-8
IUPAC Name
dichloro(phenyl)borane
Molecular Formula
C6H5BCl2
InChI Key
NCQDQONETMHUMY-UHFFFAOYSA-N
SMILES
ClB(Cl)C1=CC=CC=C1
Assay from Suppliers CofA
≥95.0%
Form
Liquid

Description

Dichlorophenylborane is widely used as a catalyst in Antiproliferative macrolide and cell migration inhibitor lactimidomycin and in enantioselective synthesis of polypeptide segments using vinylogous Mukaiyama aldol reactions and in enantioselective Diels-Alder cycloadditions after its reaction with allo-threonine derivatives. It is a stereo selective alkylative ring opening of cyclic anhydrides and in cross-metathesis reaction of amino derivatives with olefines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text