Thermo Scientific Chemicals

Benzyl azide, 94%, Thermo Scientific Chemicals

Catalog number: 032635.09
10 g, Each
Thermo Scientific Chemicals

Benzyl azide, 94%, Thermo Scientific Chemicals

Catalog number: 032635.09
10 g, Each
Quantity
Catalog number: 032635.09
also known as 032635-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
622-79-7
IUPAC Name
(azidomethyl)benzene
Molecular Formula
C7H7N3
InChI Key
UDLLFLQFQMACJB-UHFFFAOYSA-N
SMILES
[N-]=[N+]=NCC1=CC=CC=C1
Assay (GC)
≥92.0%
Refractive Index
1.5255-1.5325 @ 20?C
Appearance (Color)
Clear colorless to pale brown
Form
Liquid
Infrared spectrum
Conforms

Description

Benzyl azide is used in the synthesis of 1,2,3-triazole derivatives. It serves as a reagent used to introduce a PhCH2N group into a molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text