Thermo Scientific Chemicals

Bromophenol Blue, ACS

Catalog number: 032641.03
1 g, Each
Thermo Scientific Chemicals

Bromophenol Blue, ACS

Catalog number: 032641.03
1 g, Each
Quantity
Catalog number: 032641.03
also known as 032641-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
115-39-9
IUPAC Name
3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Formula
C19H10Br4O5S
InChI Key
UDSAIICHUKSCKT-UHFFFAOYSA-N
SMILES
OC1=C(Br)C=C(C=C1Br)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1
Comment
Visual transition interval pH 3.0 (yellow) to pH 4.6 (blue)
Comment
Clarity of solution P.T.

Description

Bromophenol blue is used as a tracking dye in electrophoresis, as a color marker in gel electrophoresis and polyacrylamide gel electrophoresis. As a biological stain, it plays an important role to stain proteins and nucleic acids. It acts an intermediate and an acid-base indicator in the pH range 3.0 to 4.6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text