Thermo Scientific Chemicals

Hydrazine dihydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: 032725.36
500 g, Each
Thermo Scientific Chemicals

Hydrazine dihydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: 032725.36
500 g, Each
Quantity
Catalog number: 032725.36
also known as 032725-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5341-61-7
IUPAC Name
hydrogen hydrazine chloride
Molecular Formula
ClH5N2
InChI Key
BIVUUOPIAYRCAP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NN
Assay (unspecified)
≥98.0%
Form
Powder or crystals or crystalline powder
Appearance (Color)
White

Description

Hydrazine dihydrochloride is used as chlorine scavenger in HCl gas streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text