Thermo Scientific Chemicals

Cyanogen bromide, Thermo Scientific Chemicals

Catalog number: 032731.22
100 g, Each
Thermo Scientific Chemicals

Cyanogen bromide, Thermo Scientific Chemicals

Catalog number: 032731.22
100 g, Each
Quantity
Catalog number: 032731.22
also known as 032731-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
506-68-3
IUPAC Name
carbononitridic bromide
Molecular Formula
CBrN
InChI Key
ATDGTVJJHBUTRL-UHFFFAOYSA-N
SMILES
BrC#N
Form
Crystals or crystalline granules or powder
Assay from Supplier's CofA
≥96.0 to ≤104.0% (Titration)
Appearance (Color)
Colorless to white

Description

Cyanogen bromide is used as a reagent in molecular biology, peptide mapping, gold extraction and amino acid sequence analysis. It acts as a coupling reagent for phosphoramidate or pyrophosphate inter-nucleotide bonds in DNA duplexes. It is also used in organic synthesis to prepare cyanamides, dicyanamides, cyanuric bromide, guanidines and hydroxylguanidines. It is involved in the von Braun reaction, it gives disubstituted cyanamides by reacting with tertiary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text