Thermo Scientific Chemicals

Copper(II) hydroxide, tech. 94%, stab., Thermo Scientific Chemicals

Catalog number: 032733.36
500 g, Each
Thermo Scientific Chemicals

Copper(II) hydroxide, tech. 94%, stab., Thermo Scientific Chemicals

Catalog number: 032733.36
500 g, Each
Quantity
Catalog number: 032733.36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20427-59-2
IUPAC Name
copper(II) hydroxide
Molecular Formula
CuH4O2
InChI Key
AEJIMXVJZFYIHN-UHFFFAOYSA-N
SMILES
O.O.[Cu]
Appearance (Color)
Pale blue
Comment
Stabilized
Assay (unspecified)
≥92.0%
Form
Powder

Description

Used as a raw material in chemical processing and catalyst manufacturing. Copper(II) hydroxide is used as ceramic colorant. In organic synthesis, it is used as a catalyst to prepare 1-((2-aminoethyl)amino)anthraquinone, 1-amino-4-((2-aminoethyl)amino)anthraquinone by reacting with 1-bromoanthraquinone or 1-amino-4-bromoanthraquinone respectively. It is involved in the synthesis of benzoic acid and octanoic acid. It is used as a precursor in the production of rayon, battery electrodes and other copper salts It is also used as a dye mordant, paper colorant, pigment and feed additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text