Manganese(II) bromide, anhydrous, 99%, H{2}O <0.3%, Thermo Scientific Chemicals
Manganese(II) bromide, anhydrous, 99%, H{2}O &lt;0.3%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) bromide, anhydrous, 99%, H{2}O <0.3%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 032736.09
also known as 032736-09
Price (USD)/ Each
63.65
Online exclusive
70.90 
Save 7.25 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
63.65
Online exclusive
70.90 
Save 7.25 (10%)
Add to cart
Manganese(II) bromide, anhydrous, 99%, H{2}O <0.3%, Thermo Scientific Chemicals
Catalog number032736.09
Price (USD)/ Each
63.65
Online exclusive
70.90 
Save 7.25 (10%)
-
Add to cart
Chemical Identifiers
CAS13446-03-2
IUPAC Namemanganese(2+) dibromide
Molecular FormulaBr2Mn
InChI KeyRJYMRRJVDRJMJW-UHFFFAOYSA-L
SMILES[Mn++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink
FormPowder
Assay from Suppliers CofA≥98.5% (Titration)
It can be used in place of palladium in the Stille reaction, which couples two carbon atoms using an organotin compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only