Thermo Scientific Chemicals

Manganese(II) bromide, anhydrous, 99%, H{2}O <0.3%, Thermo Scientific Chemicals

Catalog number: 032736.18
50 g, Each
Thermo Scientific Chemicals

Manganese(II) bromide, anhydrous, 99%, H{2}O <0.3%, Thermo Scientific Chemicals

Catalog number: 032736.18
50 g, Each
Quantity
Catalog number: 032736.18
also known as 032736-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13446-03-2
IUPAC Name
manganese(2+) dibromide
Molecular Formula
Br2Mn
InChI Key
RJYMRRJVDRJMJW-UHFFFAOYSA-L
SMILES
[Mn++].[Br-].[Br-]
Appearance (Color)
Pink
Form
Powder
Assay from Suppliers CofA
≥98.5% (Titration)

Description

It can be used in place of palladium in the Stille reaction, which couples two carbon atoms using an organotin compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text