1,4-Butanediol diacrylate, 85+%, stab. with 50-105 ppm hydroquinone, Thermo Scientific Chemicals
1,4-Butanediol diacrylate, 85+%, stab. with 50-105 ppm hydroquinone, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Butanediol diacrylate, 85+%, stab. with 50-105 ppm hydroquinone, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 032780.22
also known as 032780-22
Price (USD)/ Each
76.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.70 
Save 13.05 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
76.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.70 
Save 13.05 (15%)
Add to cart
1,4-Butanediol diacrylate, 85+%, stab. with 50-105 ppm hydroquinone, Thermo Scientific Chemicals
Catalog number032780.22
Price (USD)/ Each
76.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.70 
Save 13.05 (15%)
-
Add to cart
Chemical Identifiers
CAS1070-70-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear
Viscosity2.0 to 10.0 cps ( 25 deg C)
Assay (GC)85.0% min
Assay (HPLC)50 to 105 ppm Hydroquinone
Formviscous liquid
1,4-Butanediol diacrylate is used as a multifunctional crosslinker. It is used in drying technology for paintings, coatings, inks, resins and adhesives. It serves as an intermediate product for polymer synthesis in the chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only