Thermo Scientific Chemicals

L-Ascorbic acid 6-palmitate, 99%, Thermo Scientific Chemicals

Catalog number: 032791.14
25 g, Each
Thermo Scientific Chemicals

L-Ascorbic acid 6-palmitate, 99%, Thermo Scientific Chemicals

Catalog number: 032791.14
25 g, Each
Quantity
Catalog number: 032791.14
also known as 032791-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
137-66-6
IUPAC Name
2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate
Molecular Formula
C22H38O7
InChI Key
NULLRXHRYOXSEW-UHFFFAOYNA-N
SMILES
CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O
Assay (Titration)
> 98.5% (dried basis)
Loss on Drying
1.00% max
Sulfated Ash
0.10% max
Trace Metals, ppm (USP)

Description

L-Ascorbyl palmitate (Asc-6P) is used along with other fatty acid ascorbates such as L-ascorbyl oleate and L-ascorbyl linoleate to protect cells and membranes from oxidative damage. Fatty acid ascorbate molecules have increased solubility in non-polar (lipophilic) environments. It is applied as humectants and emollients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text