Thermo Scientific Chemicals

alpha,alpha-Dimethylphenylacetic acid, Thermo Scientific Chemicals

Catalog number: 032884.09
10 g, Each
Thermo Scientific Chemicals

alpha,alpha-Dimethylphenylacetic acid, Thermo Scientific Chemicals

Catalog number: 032884.09
10 g, Each
Quantity
Catalog number: 032884.09
also known as 032884-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
826-55-1
IUPAC Name
2-methyl-2-phenylpropanoic acid
Molecular Formula
C10H12O2
InChI Key
YYEROYLAYAVZNW-UHFFFAOYSA-N
SMILES
CC(C)(C(O)=O)C1=CC=CC=C1
Appearance (Color)
White to pale cream to pale yellow
Assay from Supplier's CofA
≥97.0%
Form
Powder or crystals or crystalline powder
Infrared spectrum
Conforms

Description

α,α-Dimethylphenylacetic acid is used in the synthesis of active pharmaceutical ingredient, fexofenadine hydrochloride, which is used as antihistamine. Further, it acts as pharmaceutical intermediates and as chemical intermediates in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text