Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Diameter (Metric) Outer | Diameter (Metric) Inner | Height (Metric) | Capacity (Metric) |
---|---|---|---|---|
032954.KF also known as 032954-KF | 40mm | 36mm | 60mm | 60 mL |
032955.KF also known as 032955-KF | 50mm | 44mm | 75mm | 110 mL |
032953.KF also known as 032953-KF | 35mm | 30mm | 50mm | 40 mL |
032952.KF also known as 032952-KF | 30mm | 26mm | 40mm | 20 mL |
032951.KF also known as 032951-KF | 20mm | 16mm | 30mm | 6 mL |