4-Methyl-1-pentene, 98+%, Thermo Scientific Chemicals
4-Methyl-1-pentene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methyl-1-pentene, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number 033029.18
also known as 033029-18
Price (USD)/ Each
140.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
140.00
Add to cart
4-Methyl-1-pentene, 98+%, Thermo Scientific Chemicals
Catalog number033029.18
Price (USD)/ Each
140.00
-
Add to cart
Specifications
Chemical Name or Material4-Methyl-1-pentene
CAS691-37-2
Health Hazard 1H225
Health Hazard 2GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
View more
4-Methyl-1-pentene is used as a monomer for olefin polymerisation. The resulting polymer is poly(4-methyl-1-pentene). It can also be used to produce 1,2-diiodo-4-methyl-pentane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only