Al-23 Boat;L1 (mm), 87;L2 (mm), 72;B1 (mm), 15;B2 (mm), 13;H (mm), 8;W (mm), 2.5;Vol, 3.4, Thermo Scientific Chemicals
Al-23 Boat;L1 (mm), 87;L2 (mm), 72;B1 (mm), 15;B2 (mm), 13;H (mm), 8;W (mm), 2.5;Vol, 3.4, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Al-23 Boat;L1 (mm), 87;L2 (mm), 72;B1 (mm), 15;B2 (mm), 13;H (mm), 8;W (mm), 2.5;Vol, 3.4, Thermo Scientific Chemicals
Have Questions?
Quantity:
1 Pc.
Catalog number 033175.KF
Price (USD)/ Each
63.80
-
Add to cart
Quantity:
1 Pc.
Request bulk or custom format
Price (USD)/ Each
63.80
Add to cart
Al-23 Boat;L1 (mm), 87;L2 (mm), 72;B1 (mm), 15;B2 (mm), 13;H (mm), 8;W (mm), 2.5;Vol, 3.4, Thermo Scientific Chemicals
Catalog number033175.KF
Price (USD)/ Each
63.80
-
Add to cart
Specifications
Chemical Name or MaterialAl-23 Boat
Recommended StorageAmbient temperatures
TSCAU
Quantity1 Pc.
FormBoat