Thermo Scientific Chemicals

Phenol, ACS, 99+%, stab., Thermo Scientific Chemicals

Catalog number: 033213.36
500 g, Each
Thermo Scientific Chemicals

Phenol, ACS, 99+%, stab., Thermo Scientific Chemicals

Catalog number: 033213.36
500 g, Each
Quantity
Catalog number: 033213.36
also known as 033213-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-95-2
IUPAC Name
phenol
Molecular Formula
C6H6O
InChI Key
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1
Clarity
Clarity of solution Passes test
Residue on Evaporation
0.05%, max
Assay (unspecified)
Freezing point Not below 40.5 degC (dry basis)
Water Content (Karl Fischer Titration)
0.5%, max
Assay (Iodometric Titration)
>/= 99.0%

Description

Phenol is used as a precursor for cyclohexanone, plastics, nonionic detergents and pharmaceutical drugs like aspirin. It acts as an anesthetic in chloraseptic. It reacts with acetone to get bisphenol-A which is used in the synthesis of poly carbonates and epoxide resins. It is also used in the manufacture of synthetic resins, dyes, pharmaceuticals, synthetic tanning agents, perfumes, lubricating oils and solvents. In molecular biology, it is used in the extraction of nucleic acid from tissues by using the liquid/liquid phenol-chloroform extraction technique. It is an active component of paint strippers which are used for the removal of epoxy and polyurethane. It is also used in the preparation of cosmetics, hair colors and skin lightening preparations. In the field of medicine, its spray is useful in helping sore throat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text