Bismuth carbonate oxide, 98.5% min, Thermo Scientific Chemicals
Bismuth carbonate oxide, 98.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth carbonate oxide, 98.5% min, Thermo Scientific Chemicals

Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazes
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 033216.36
also known as 033216-36
Price (USD)/ Each
234.65
Online exclusive
261.00 
Save 26.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
234.65
Online exclusive
261.00 
Save 26.35 (10%)
Add to cart
Bismuth carbonate oxide, 98.5% min, Thermo Scientific Chemicals
Catalog number033216.36
Price (USD)/ Each
234.65
Online exclusive
261.00 
Save 26.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5892-10-4
IUPAC Name({[(oxobismuthanyl)oxy]carbonyl}oxy)bismuthanone
Molecular FormulaCBi2O5
InChI KeyMGLUJXPJRXTKJM-UHFFFAOYSA-L
SMILESO=[Bi]OC(=O)O[Bi]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.5% min
Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazesBismuth carbonate oxide is used in ceramic glazes and pearly coatings on plastics. It is also used as an antacid and anti-diarrheal in medicinal chemistry. Further, it is used as a filler in radiopaque catheters. It is also employed in fireworks and to make Dragon's eggs. In addition to this, it is made into nanotube arrays that exhibit antibacterial properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only