Citric acid diammonium salt, 99+%, Thermo Scientific Chemicals
Citric acid diammonium salt, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Citric acid diammonium salt, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 033221.22
also known as 033221-22
Price (USD)/ Each
44.60
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
44.60
Add to cart
Citric acid diammonium salt, 99+%, Thermo Scientific Chemicals
Catalog number033221.22
Price (USD)/ Each
44.60
-
Add to cart
Chemical Identifiers
CAS3012-65-5
IUPAC Namediammonium 3-carboxy-3-hydroxypentanedioate
Molecular FormulaC6H14N2O7
InChI KeyYXVFQADLFFNVDS-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Titration)> 99.0%
Residue on ignition0.01%, max
Citric acid diammonium salt is used in pharmaceuticals, rust proofing, cotton printing and plasticizers. It is used as a reagent for the determination of phosphate in fertilizers, and as a buffering agent, chelating agent and source of nitrogen in chicken food supplements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only