Tripotassium citrate monohydrate, 99+%, Thermo Scientific Chemicals
Tripotassium citrate monohydrate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tripotassium citrate monohydrate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 033222.36
also known as 033222-36
Price (USD)/ Each
36.65
Online exclusive
41.10 
Save 4.45 (11%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
36.65
Online exclusive
41.10 
Save 4.45 (11%)
Add to cart
Tripotassium citrate monohydrate, 99+%, Thermo Scientific Chemicals
Catalog number033222.36
Price (USD)/ Each
36.65
Online exclusive
41.10 
Save 4.45 (11%)
-
Add to cart
Chemical Identifiers
CAS6100-05-6
IUPAC Nametripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
Molecular FormulaC6H7K3O8
InChI KeyPJAHUDTUZRZBKM-UHFFFAOYSA-K
SMILESO.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Heavy Metals≤5.0 ppm
Assay from Supplier's CofA≥99.0%
Tripotassium citrate hydrate is used in food and beverage industries. It finds application as buffering, sequestering or emulsifying agent. It is also used as a potassium source in pharmaceuticals and an active ingredient to treat urinary duct stones. It is added to oral care products to reduce the teeth sensitivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only