Thermo Scientific Chemicals

Tripotassium citrate monohydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 033222.A3
2 kg, Each
Thermo Scientific Chemicals

Tripotassium citrate monohydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 033222.A3
2 kg, Each
Quantity
Catalog number: 033222.A3
also known as 033222-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6100-05-6
IUPAC Name
tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
Molecular Formula
C6H7K3O8
InChI Key
PJAHUDTUZRZBKM-UHFFFAOYSA-K
SMILES
O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Heavy Metals
≤5.0 ppm
Assay from Supplier's CofA
≥99.0%

Description

Tripotassium citrate hydrate is used in food and beverage industries. It finds application as buffering, sequestering or emulsifying agent. It is also used as a potassium source in pharmaceuticals and an active ingredient to treat urinary duct stones. It is added to oral care products to reduce the teeth sensitivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text