Thermo Scientific Chemicals

Diethyl ether, ACS, 98% min, stab. with 0.001% BHT and 3% ethanol, Thermo Scientific Chemicals

Catalog number: 033224.62
6 x 500 g, Each
Thermo Scientific Chemicals

Diethyl ether, ACS, 98% min, stab. with 0.001% BHT and 3% ethanol, Thermo Scientific Chemicals

Catalog number: 033224.62
6 x 500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 033224.62
also known as 033224-62
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60-29-7
IUPAC Name
ethoxyethane
Molecular Formula
C4H10O
InChI Key
RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILES
CCOCC
Color (APHA)
10, max
Assay (GC)
98.0% min
Free acid (titration)
0.0002 meq/g, max
Peroxide content
1 ppm ( as H2O2), max
Residue on Evaporation
0.001% max

Description

Widely used as a solvent. Dissolves P, S, Br2 and I2. It is a widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text