Cadmium oxide, 98.9%, Thermo Scientific Chemicals
Cadmium oxide, 98.9%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium oxide, 98.9%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 033235.30
also known as 033235-30
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.80 
Save 7.15 (15%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.80 
Save 7.15 (15%)
Add to cart
Cadmium oxide, 98.9%, Thermo Scientific Chemicals
Catalog number033235.30
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.80 
Save 7.15 (15%)
-
Add to cart
Chemical Identifiers
CAS1306-19-0
IUPAC Namecadmium(2+) oxidandiide
Molecular FormulaCdO
InChI KeyCFEAAQFZALKQPA-UHFFFAOYSA-N
SMILES[O--].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.9% min
Cadmium oxide is used as an electroplating chemical and in the manufacture of cadmium electrodes. It is a component of silver alloys, phosphors, semiconductors, glass and ceramic glazes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only