Thermo Scientific Chemicals

Cadmium oxide, 98.9%, Thermo Scientific Chemicals

Catalog number: 033235.A1
1 kg, Each
Thermo Scientific Chemicals

Cadmium oxide, 98.9%, Thermo Scientific Chemicals

Catalog number: 033235.A1
1 kg, Each
Quantity
Catalog number: 033235.A1
also known as 033235-A1
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1306-19-0
IUPAC Name
cadmium(2+) oxidandiide
Molecular Formula
CdO
InChI Key
CFEAAQFZALKQPA-UHFFFAOYSA-N
SMILES
[O--].[Cd++]
Assay (unspecified)
98.9% min

Description

Cadmium oxide is used as an electroplating chemical and in the manufacture of cadmium electrodes. It is a component of silver alloys, phosphors, semiconductors, glass and ceramic glazes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text