Thermo Scientific Chemicals

Sodium L-(+)-tartrate dihydrate, ACS, 99.0-101.0%, Thermo Scientific Chemicals

Catalog number: 033242.22
100 g, Each
Thermo Scientific Chemicals

Sodium L-(+)-tartrate dihydrate, ACS, 99.0-101.0%, Thermo Scientific Chemicals

Catalog number: 033242.22
100 g, Each
Quantity
Catalog number: 033242.22
also known as 033242-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6106-24-7
IUPAC Name
disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
Molecular Formula
C4H8Na2O8
InChI Key
FGJLAJMGHXGFDE-UDMGOBQRNA-L
SMILES
O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
Impurity content
Cl 5ppm
Trace Metals
(as Pb) 5ppm
Assay (unspecified)
99.0-101.0% C₄H₄Na₂O₆?2H₂O
Impurity content
Fe 0.001%
Loss on Drying
at 150? 15.61-15.71%

Description

Sodium L-(+)-tartrate is used as an emulsifier, as a food additives, binding agent in food products like jellies, margarine and sausage casings. It is utilized as a common primary standard for Karl Fischer titration to measure the water content. It is also used as buffers for molecular biology and cell culture applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text