Thermo Scientific Chemicals

Acetaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 033244.AE
100 mL, Each
Thermo Scientific Chemicals

Acetaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 033244.AE
100 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 033244.AE
also known as 033244-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-07-0
IUPAC Name
acetaldehyde
Molecular Formula
C2H4O
InChI Key
IKHGUXGNUITLKF-UHFFFAOYSA-N
SMILES
CC=O
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%

Description

Acetaldehyde is used in the production of drugs, perfumes, explosives, lacquers and varnishes, photographic chemicals, phenolic and urea resins, rubber accelerators and antioxidants. It is used as an intermediate for the synthesis of acetic acid, ethyl acetate, acetic anhydride, pentaerythritol, peracetic acid, chloral and glyoxal. As a solvent, it is used in rubber production, tanning of leather and in the paper industry. It is used as a fruit and fish preservative, as a flavoring agent, as a denaturant for alcohol and in composition of fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text