Acetaldehyde, 99%, Thermo Scientific Chemicals
Acetaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetaldehyde, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 500 mL
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 033244.AY
also known as 033244-AY
Price (USD)/ Each
519.65
Online exclusive
577.00 
Save 57.35 (10%)
-
Add to cart
Quantity:
4 x 500 mL
Request bulk or custom format
Price (USD)/ Each
519.65
Online exclusive
577.00 
Save 57.35 (10%)
Add to cart
Acetaldehyde, 99%, Thermo Scientific Chemicals
Catalog number033244.AY
Price (USD)/ Each
519.65
Online exclusive
577.00 
Save 57.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-07-0
IUPAC Nameacetaldehyde
Molecular FormulaC2H4O
InChI KeyIKHGUXGNUITLKF-UHFFFAOYSA-N
SMILESCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Acetaldehyde is used in the production of drugs, perfumes, explosives, lacquers and varnishes, photographic chemicals, phenolic and urea resins, rubber accelerators and antioxidants. It is used as an intermediate for the synthesis of acetic acid, ethyl acetate, acetic anhydride, pentaerythritol, peracetic acid, chloral and glyoxal. As a solvent, it is used in rubber production, tanning of leather and in the paper industry. It is used as a fruit and fish preservative, as a flavoring agent, as a denaturant for alcohol and in composition of fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only