Boric acid, ACS, 99.5% min, Thermo Scientific Chemicals
Boric acid, ACS, 99.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boric acid, ACS, 99.5% min, Thermo Scientific Chemicals

In hardening steel, preservatives, manufacturing of cements, porcelain, enamels, glass, borates
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 033253.A3
also known as 033253-A3
Price (USD)/ Each
117.65
Online exclusive
131.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
117.65
Online exclusive
131.00 
Save 13.35 (10%)
Add to cart
Boric acid, ACS, 99.5% min, Thermo Scientific Chemicals
Catalog number033253.A3
Price (USD)/ Each
117.65
Online exclusive
131.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10043-35-3
IUPAC Nameboric acid
Molecular FormulaBH3O3
InChI KeyKGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILESOB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCa 0.005%
Impurity contentFe 0.001%
Impurity contentInsoluble in methanol 0.005%
Impurity contentSO₄ 0.01%
Trace Metals(as Pb) 0.001%
View more
In hardening steel, preservatives, manufacturing of cements, porcelain, enamels, glass, borates, artificial gems, in nickeling baths, painting, photographyIt is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is too employed in leather manufacture, electroplating, and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only